siRNA Details
| Gene Symbol: ITGA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316312.1 | NP_001303241.1 | 5 | 1660 | ||
| GenBank mRNA | BC055419.1 | - | 5 | 1352 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR608655.1 | - | 1 | 815 | Chr. 2 - 181475346 - 181476192 [+] on Build GRCh38 | |

ITGA4
Genomic Map