siRNA Details
| Gene Symbol: GHET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130107.1 | 1 | 1852 | |||
| GenBank mRNA | AK123072.1 | - | 1 | 1790 | ||
| BC037977.1 | - | 1 | 1761 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GHET1| Gene Symbol: GHET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130107.1 | 1 | 1852 | |||
| GenBank mRNA | AK123072.1 | - | 1 | 1790 | ||
| BC037977.1 | - | 1 | 1761 | |||