siRNA Details
| Gene Symbol: ZNF141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011513562.2 | XP_011511864.1 | 1 | 1006 | ||
| XM_017008591.1 | XP_016864080.1 | 1 | 1002 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK123078.1 | - | 1 | 63 | Chr. 4 - 342263 - 344792 [+] on Build GRCh38 | |

ZNF141
Genomic Map