siRNA Details
| Gene Symbol: LOC401442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134292.1 | 1 | 1018 | |||
| GenBank mRNA | BC028401.1 | - | 1 | 832 | ||
| BC067903.1 | - | 1 | 1018 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401442| Gene Symbol: LOC401442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134292.1 | 1 | 1018 | |||
| GenBank mRNA | BC028401.1 | - | 1 | 832 | ||
| BC067903.1 | - | 1 | 1018 | |||