siRNA Details
| Gene Symbol: LOC105378146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136248.1 | 2 | 2489 | |||
| NR_136249.1 | 3 | 2104 | ||||
| GenBank mRNA | AY726573.1 | - | 1 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378146| Gene Symbol: LOC105378146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136248.1 | 2 | 2489 | |||
| NR_136249.1 | 3 | 2104 | ||||
| GenBank mRNA | AY726573.1 | - | 1 | 92 | ||