siRNA Details
Gene Symbol: LOC105378146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136248.1 | 2 | 2563 | |||
NR_136249.1 | 3 | 2178 | ||||
GenBank mRNA | AY726573.1 | - | 1 | 166 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105378146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136248.1 | 2 | 2563 | |||
NR_136249.1 | 3 | 2178 | ||||
GenBank mRNA | AY726573.1 | - | 1 | 166 |