siRNA Details
| Gene Symbol: LOC105378146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136248.1 | 2 | 3091 | |||
| NR_136249.1 | 3 | 2706 | ||||
| GenBank mRNA | AY726573.1 | - | 1 | 694 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB521434.1 | - | 1 | 403 | Chr. 6 - 168960408 - 168960876 [-] on Build GRCh38 | |

LOC105378146
Genomic Map