siRNA Details
| Gene Symbol: CREB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK298121.1 | - | 3 | 629 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK021493.1 | - | 1 | 342 | Chr. 7 - 28724576 - 28725645 [+] on Build GRCh38 | |

CREB5
Genomic Map