siRNA Details
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011539031.1 | XP_011537333.1 | 1 | 2667 | ||
| XM_011539032.1 | XP_011537334.1 | 1 | 2667 | |||
| GenBank mRNA | AK124275.1 | - | 1 | 2245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLEC| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011539031.1 | XP_011537333.1 | 1 | 2667 | ||
| XM_011539032.1 | XP_011537334.1 | 1 | 2667 | |||
| GenBank mRNA | AK124275.1 | - | 1 | 2245 | ||