siRNA Details
| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000855.2 | NP_000846.1 | 8 | 12608 | ||
| NM_001256424.1 | NP_001243353.1 | 9 | 12701 | |||
| GenBank mRNA | AK001875.1 | - | 1 | 1125 | ||
| AK022090.1 | - | 1 | 871 | |||

GUCY1A2
Genomic Map