siRNA Details
| Gene Symbol: DPYS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516903.2 | XP_011515205.1 | 10 | 2323 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK055858.1 | - | 1 | 34 | Chr. 8 - 104383619 - 104387788 [-] on Build GRCh38 | |

DPYS
Genomic Map