siRNA Details
| Gene Symbol: LOC100128035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY358772.1 | - | 1 | 565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100128035| Gene Symbol: LOC100128035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY358772.1 | - | 1 | 565 | ||