siRNA Details
| Gene Symbol: LINC01545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046101.1 | 3 | 222 | |||
| GenBank mRNA | BC038573.2 | - | 3 | 222 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG254667.1 | - | 3 | 232 | Chr. X - 46887387 - 46899704 [-] on Build GRCh38 | |

LINC01545
Genomic Map