siRNA Details
Gene Symbol: LINC01545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046101.1 | 4 | 458 | |||
GenBank mRNA | BC038573.2 | - | 4 | 458 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG254667.1 | - | 4 | 468 | Chr. X - 46887387 - 46899704 [-] on Build GRCh38 |
