siRNA Details
| Gene Symbol: LOC339298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040034.1 | 3, 4 | 2074 | |||
| GenBank mRNA | BC043002.2 | - | 3, 4 | 2065 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000584677.1 | - | 3, 4 | 2074 | Chr. 18 - 76122998 - 76145255 [+] on Build GRCh38 | |

LOC339298
Genomic Map