siRNA Details
| Gene Symbol: PDE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258312.1 | NP_001245241.1 | 16 | 3631 | ||
| NM_005019.4 | NP_005010.2 | 15 | 3646 | |||
| XM_017004295.1 | XP_016859784.1 | 16 | 3651 | |||
| XM_017004296.1 | XP_016859785.1 | 16 | 3573 | |||
| XM_017004297.1 | XP_016859786.1 | 16 | 3514 | |||
| XM_017004298.1 | XP_016859787.1 | 16 | 3696 | |||
| XM_017004302.1 | XP_016859791.1 | 15 | 3778 | |||
| GenBank mRNA | BC047057.1 | - | 1 | 321 | ||

PDE1A
Genomic Map