siRNA Details
| Gene Symbol: LOC101930370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134241.1 | 4 | 1497 | |||
| GenBank mRNA | BC043556.1 | - | 4 | 1000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101930370| Gene Symbol: LOC101930370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134241.1 | 4 | 1497 | |||
| GenBank mRNA | BC043556.1 | - | 4 | 1000 | ||