siRNA Details
| Gene Symbol: LOC101930370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134241.1 | 4 | 2145 | |||
| GenBank mRNA | BC043556.1 | - | 4 | 1648 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ638916.1 | - | 1 | 486 | Chr. 4 - 173144316 - 173144888 [-] on Build GRCh38 | |

LOC101930370
Genomic Map