siRNA Details
| Gene Symbol: LINC01395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120582.1 | 1 | 41 | |||
| GenBank mRNA | BC038579.2 | - | 1 | 41 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI859347.1 | - | 2 | 60 | Chr. 11 - 129614325 - 129681152 [-] on Build GRCh38 | |

LINC01395
Genomic Map