siRNA Details
| Gene Symbol: LOC105369187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135033.1 | 1 | 188 | |||
| GenBank mRNA | BC043551.1 | - | 1 | 188 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI831076.1 | - | 1 | 188 | Chr. 12 - 65544273 - 65557311 [-] on Build GRCh38 | |

LOC105369187
Genomic Map