siRNA Details
| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253384.1 | NP_001240313.1 | 5 | 1945 | ||
| GenBank mRNA | BC081565.1 | - | 5 | 1610 | 5458912 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK055501.1 | - | 1 | 548 | Chr. 3 - 23922255 - 23924373 [+] on Build GRCh38 | |

RPL15
Genomic Map