siRNA Details
| Gene Symbol: LOC101927331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135589.1 | 2 | 222 | |||
| GenBank mRNA | BC014368.1 | - | 2 | 223 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000449772.1 | - | 2 | 199 | Chr. 2 - 104412227 - 104414008 [-] on Build GRCh38 | |

LOC101927331
Genomic Map