siRNA Details
| Gene Symbol: SNAP25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040710.1 | 5 | 1484 | |||
| GenBank mRNA | AK124961.1 | - | 5 | 1481 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR609457.1 | - | 1 | 1365 | Chr. 20 - 10023856 - 10026651 [-] on Build GRCh38 | |

SNAP25-AS1
Genomic Map