siRNA Details
| Gene Symbol: FAM172A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005272111.4 | XP_005272168.1 | 8 | 4056 | ||
| XM_017009962.1 | XP_016865451.1 | 7 | 3922 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022140.1 | - | 1 | 700 | Chr. 5 - 93860669 - 93863825 [-] on Build GRCh38 | |

FAM172A
Genomic Map