siRNA Details
| Gene Symbol: IFI44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011540516.2 | XP_011538818.1 | 8 | 2651 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX537508.1 | - | 2 | 1842 | Chr. 1 - 78358729 - 78664078 [+] on Build GRCh38 | |

IFI44
Genomic Map