siRNA Details
| Gene Symbol: MZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB209481.1 | - | 2 | 3052 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126582.1 | - | 1 | 375 | Chr. 19 - 58566920 - 58568750 [-] on Build GRCh38 | |

MZF1
Genomic Map