siRNA Details
| Gene Symbol: MANEA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047502.1 | 1 | 1721 | |||
| GenBank mRNA | BC047582.1 | - | 1 | 1720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MANEA-AS1| Gene Symbol: MANEA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047502.1 | 1 | 1721 | |||
| GenBank mRNA | BC047582.1 | - | 1 | 1720 | ||