siRNA Details
| Gene Symbol: LINC01480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110724.1 | 3 | 330 | |||
| GenBank mRNA | AK123333.1 | - | 1 | 1577 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01480| Gene Symbol: LINC01480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110724.1 | 3 | 330 | |||
| GenBank mRNA | AK123333.1 | - | 1 | 1577 | ||