siRNA Details
| Gene Symbol: PPP2R2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177591.1 | NP_001171062.1 | 10 | 3126 | ||
| NM_002717.3 | NP_002708.1 | 10 | 3394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2R2A| Gene Symbol: PPP2R2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177591.1 | NP_001171062.1 | 10 | 3126 | ||
| NM_002717.3 | NP_002708.1 | 10 | 3394 | |||