siRNA Details
| Gene Symbol: PPP2R2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177591.1 | NP_001171062.1 | 10 | 2509 | ||
| NM_002717.3 | NP_002708.1 | 10 | 2777 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL050206.1 | - | 1 | 190 | Chr. 8 - 26371375 - 26372175 [+] on Build GRCh38 | |

PPP2R2A
Genomic Map