siRNA Details
| Gene Symbol: CYYR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135515.1 | 4 | 2298 | |||
| GenBank mRNA | AK125338.1 | - | 4 | 2298 | ||
| JF693915.1 | - | 4 | 2135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYYR1-AS1| Gene Symbol: CYYR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135515.1 | 4 | 2298 | |||
| GenBank mRNA | AK125338.1 | - | 4 | 2298 | ||
| JF693915.1 | - | 4 | 2135 | |||