siRNA Details
| Gene Symbol: LZIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316973.1 | NP_001303902.1 | 8 | 4419 | ||
| NM_001316974.1 | NP_001303903.1 | 8 | 4362 | |||
| NM_001316976.1 | NP_001303905.1 | 6 | 4241 | |||
| NM_032368.4 | NP_115744.2 | 8 | 4519 | |||
| XM_005263509.2 | XP_005263566.1 | 6 | 4236 | |||
| XM_017002546.1 | XP_016858035.1 | 8 | 4265 | |||
| XM_017002547.1 | XP_016858036.1 | 8 | 4285 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR617316.1 | - | 1 | 611 | Chr. 1 - 9926393 - 9927409 [-] on Build GRCh38 | |

LZIC
Genomic Map