siRNA Details
| Gene Symbol: NAV2-AS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033989.1 | 3 | 487 | |||
| GenBank mRNA | AK023501.1 | - | 3 | 487 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU135729.1 | - | 3 | 487 | Chr. 11 - 19511403 - 19524078 [-] on Build GRCh38 | |

NAV2-AS4
Genomic Map