siRNA Details
| Gene Symbol: GRAMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530013.2 | XP_011528315.2 | 18 | 1753 | ||
| GenBank mRNA | BC033170.1 | - | 1 | 733 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRAMD4| Gene Symbol: GRAMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530013.2 | XP_011528315.2 | 18 | 1753 | ||
| GenBank mRNA | BC033170.1 | - | 1 | 733 | ||