siRNA Details
Gene Symbol: GRAMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530013.2 | XP_011528315.2 | 18 | 2047 | ||
GenBank mRNA | BC033170.1 | - | 1 | 1027 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GRAMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530013.2 | XP_011528315.2 | 18 | 2047 | ||
GenBank mRNA | BC033170.1 | - | 1 | 1027 |