siRNA Details
Gene Symbol: LOC101929645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109948.1 | 3 | 1599 | |||
GenBank mRNA | AL137617.1 | - | 3 | 1599 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109948.1 | 3 | 1599 | |||
GenBank mRNA | AL137617.1 | - | 3 | 1599 |