siRNA Details
| Gene Symbol: LOC101929645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109948.1 | 3 | 1977 | |||
| GenBank mRNA | AL137617.1 | - | 3 | 1977 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929645| Gene Symbol: LOC101929645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109948.1 | 3 | 1977 | |||
| GenBank mRNA | AL137617.1 | - | 3 | 1977 | ||