siRNA Details
| Gene Symbol: LOC102606465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109849.1 | 4 | 702 | |||
| GenBank mRNA | AK021551.1 | - | 1 | 1025 | ||
| AL529452.3 | - | 4 | 629 | |||
| BC045807.2 | - | 5 | 680 | |||
| BX538249.1 | - | 7 | 1407 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102606465| Gene Symbol: LOC102606465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109849.1 | 4 | 702 | |||
| GenBank mRNA | AK021551.1 | - | 1 | 1025 | ||
| AL529452.3 | - | 4 | 629 | |||
| BC045807.2 | - | 5 | 680 | |||
| BX538249.1 | - | 7 | 1407 | |||