siRNA Details
| Gene Symbol: LOC101928495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110409.1 | 2 | 164 | |||
| GenBank mRNA | BC013926.1 | - | 2 | 174 | ||
| Gene Symbol: GAP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF279774.1 | - | 1, 2 | 222 | ||
| Gene Symbol: LOC105378763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF279783.1 | - | 1, 2 | 232 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE564886.1 | - | 150 | Chr. - - [-] on Build | ||

LOC101928495
Genomic Map