siRNA Details
| Gene Symbol: SUGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 3045 | ||
| XM_011534893.2 | XP_011533195.1 | 13 | 2763 | |||
| XM_017020365.1 | XP_016875854.1 | 14 | 2935 | |||
| XM_017020366.1 | XP_016875855.1 | 14 | 2810 | |||
| XM_017020367.1 | XP_016875856.1 | 14 | 2844 | |||
| XM_017020368.1 | XP_016875857.1 | 14 | 2856 | |||

SUGT1
Genomic Map