siRNA Details
| Gene Symbol: CCDC134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005261748.3 | XP_005261805.1 | 8 | 3771 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124515.1 | - | 1 | 1082 | Chr. 22 - 41827573 - 41832162 [+] on Build GRCh38 | |

CCDC134
Genomic Map