siRNA Details
| Gene Symbol: MARCH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282807.1 | NP_001269736.1 | 1 | 93 | ||
| GenBank mRNA | AK302347.1 | - | 1 | 42 | ||
| AK310238.1 | - | 1 | 50 | |||
| AK316382.1 | - | 1 | 50 | |||
| BI462287.1 | - | 98 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK023400.1 | - | 1 | 1013 | Chr. 2 - 159732960 - 159734817 [+] on Build GRCh38 | |

MARCH7
Genomic Map