siRNA Details
| Gene Symbol: KXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648739.1 | - | 5 | 2376 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY336111.1 | - | 1 | 188 | Chr. 19 - 18559118 - 18559548 [-] on Build GRCh38 | |

KXD1
Genomic Map