siRNA Details
| Gene Symbol: BRD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134296.1 | 15 | 2153 | |||
| GenBank mRNA | BC007217.1 | - | 16 | 1943 | ||
| BX648703.1 | - | 15 | 2155 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX649024.1 | - | 1 | 128 | Chr. 2 - 113762423 - 113766911 [+] on Build GRCh38 | |

BRD9
Genomic Map