siRNA Details
| Gene Symbol: LOC101243545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102265.2 | 1 | 1097 | |||
| GenBank mRNA | AK094885.1 | - | 1 | 1099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101243545| Gene Symbol: LOC101243545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102265.2 | 1 | 1097 | |||
| GenBank mRNA | AK094885.1 | - | 1 | 1099 | ||