siRNA Details
| Gene Symbol: PIAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324046.1 | NP_001310975.1 | 16 | 11243 | ||
| NM_001324047.1 | NP_001310976.1 | 15 | 11183 | |||
| NM_001324048.1 | NP_001310977.1 | 16 | 11269 | |||
| NM_001324049.1 | NP_001310978.1 | 17 | 11392 | |||
| NM_004671.4 | NP_004662.2 | 14 | 11071 | |||
| NR_136684.1 | 13 | 10986 | ||||
| GenBank mRNA | AF086240.1 | - | 1 | 480 | ||

PIAS2
Genomic Map