siRNA Details
| Gene Symbol: LOC101927735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110058.1 | 5 | 1054 | |||
| GenBank mRNA | BC040912.1 | - | 5 | 1054 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000542578.1 | - | 5 | 1054 | Chr. 12 - 129208601 - 129212662 [+] on Build GRCh38 | |

LOC101927735
Genomic Map