siRNA Details
| Gene Symbol: LOC101929460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134622.1 | 2 | 839 | |||
| GenBank mRNA | BC040917.1 | - | 2 | 839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929460| Gene Symbol: LOC101929460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134622.1 | 2 | 839 | |||
| GenBank mRNA | BC040917.1 | - | 2 | 839 | ||