siRNA Details
| Gene Symbol: NSUN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011519384.1 | XP_011517686.1 | 2 | 304 | ||
| GenBank mRNA | AL698095.1 | - | 2 | 299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NSUN6| Gene Symbol: NSUN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011519384.1 | XP_011517686.1 | 2 | 304 | ||
| GenBank mRNA | AL698095.1 | - | 2 | 299 | ||