siRNA Details
Gene Symbol: NSUN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519384.1 | XP_011517686.1 | 2 | 109 | ||
GenBank mRNA | AL698095.1 | - | 2 | 104 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NSUN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519384.1 | XP_011517686.1 | 2 | 109 | ||
GenBank mRNA | AL698095.1 | - | 2 | 104 |